821854
TOYOPEARL GigaCap® Q-650M Bulk Media
bottle of 100 mL, phase ammonium ion, quaternary, 75 μm particle size
Synonym(s):
TOYOPEARL GigaCap® Q-650 Anion Exchange Media
About This Item
Recommended Products
product line
TOYOPEARL®
form
slurry
feature
counter ion Cl-
packaging
bottle of 100 mL
manufacturer/tradename
Tosoh 21854
parameter
3 bar max. pressure
technique(s)
LPLC: suitable
matrix
hydroxylated methacrylic polymer
matrix active group
ammonium ion, quaternary phase
particle size
75 μm
operating pH
3-13
capacity
>162 g/L adsorption capacity (BSA)
separation technique
anion exchange
Looking for similar products? Visit Product Comparison Guide
General description
Toyopearl GigaCap Q-650M resin exhibits typical dynamic binding capacities in the vicinity of 175 mg/mL for BSA and 49 mg/mL for thyroglobulin, whereas, Toyopearl GigaCap Q-650S has dynamic binding capacities approaching 190 g/L for bovine serum albumin.
Toyopearl GigaCap Q-650 resin maintains high capacity at increased linear velocities, has good pressure-flow characteristics and will withstand back pressures up to 3 bar. The high capacity, combined with the 35 μm mean particle size, makes Toyopearl GigaCap Q-650S ideal for high resolution applications.
In addition to the high DBC and good pressure flow characteristics, Toyopearl GigaCap Q-650 resin displays excellent binding and elution kinetics, which results in a more concentrated protein fraction collected during the elution step.
*specifically designed for packed bed use
Specifications
Physical form
Legal Information
related product
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Liq. 3
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
104.0 °F - closed cup
Flash Point(C)
40 °C - closed cup
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service