Skip to Content
Merck
All Photos(1)

Key Documents

58703

PTFE Tubing

L × O.D. × I.D. 10 ft × 1/8 in. × 0.063 in.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700

packaging

pkg of 1

parameter

500 psi max. pressure

L × O.D. × I.D.

10 ft × 1/8 in. × 0.063 in.

Looking for similar products? Visit Product Comparison Guide

General description

Economical, flexible PTFE tubing is ideal for use at pressures up to 500 psi (35 kg/cm2). Use in automatic analyzer, postcolumn reaction, preparative scale systems, in-stream sampling devices, and when monitoring physiologically important compounds.

The maximum recommended operating temperature for PTFE is 200 °C, but short term exposure to higher temperatures seldom causes damage.
Polytetrafluoroethylene (PTFE) is a fluoropolymer which is the most lubricious polymer with low co-efficient of friction. PTFE tubing is chosen due to its excellent hydrophobicity, chemical inertness, and ability to withstand high pressures. It is suitable for protecting the optical fibres.

Application

PTFE tubing may be used to connect the gas lines to the ports during Gas Chromatography-mass spectrometry analysis of volatile organic compounds from potato tubers inoculated with Phytophthora infestans or Fusarium coeruleum.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gas chromatography-mass spectrometry analyses of volatile organic compounds from potato tubers inoculated with Phytophthora infestans or Fusarium coeruleum.
De Lacy Costello, B. P. J., et al.
Plant Pathology, 50, 489-496 (2001)
K. T. V. Grattan, B.T. Meggitt
Optical Fiber Sensor Technology: Applications and systems, 3, 36-36 (1996)
Buddy D. Ratner
Biomaterials Science: An Introduction to Materials in Medicine, 93-93 (2012)
Tania Nolan, Stephen A.Bustin
PCR Technology, 164-164 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service