Skip to Content
Merck
All Photos(1)

Documents

47733-U

Supelco

2-Hexanone

analytical standard

Synonym(s):

Butyl methyl ketone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3COCH3
CAS Number:
Molecular Weight:
100.16
Beilstein:
1737676
EC Number:
MDL number:
UNSPSC Code:
77101502
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Agency

ASTM® D5580

vapor pressure

10 mmHg ( 39 °C)

CofA

current certificate can be downloaded

packaging

pkg of 5 mL

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.401 (lit.)

bp

127 °C (lit.)

mp

−57 °C (lit.)

density

0.812 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care
petroleum

format

neat

storage temp.

2-30°C

SMILES string

CCCCC(C)=O

InChI

1S/C6H12O/c1-3-4-5-6(2)7/h3-5H2,1-2H3

InChI key

QQZOPKMRPOGIEB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Hexanone is a colorless to pale yellow liquid with sharp odor belonging to the class of ketones. It is obtained as a waste product of coal gasification, oil shale operations and wood pulping. It is widely used as a solvent in industry.

Application

2-Hexanone may be used as an analytical reference standard for the quantification of the analyte in biological samples using liquid chromatography−tandem mass spectrometry (LC−MS−MS).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 3 - Repr. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

73.4 °F - closed cup

Flash Point(C)

23 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Application of liquid chromatography-mass spectrometry to biomonitoring of exposure to industrial chemicals
Manini P, et al.
Toxicology Letters, 162(2-3), 202-210 (2006)
Handbook of Chemicals and Safety (2016)
J W Griffin
Neurobehavioral toxicology and teratology, 3(4), 437-444 (1981-01-01)
The neurotoxicity of hexacarbon solvents has become apparent only recently, but an extensive literature has already developed as a result of the clinical and epidemiological implications of the human disease, and the advantages of small animal models of hexacarbon neurotoxicity
Y Lolin
Human toxicology, 8(4), 293-300 (1989-07-01)
1. The main neurological disorders associated with chronic VSA are peripheral neuropathy, cerebellar disease, chronic encephalopathy and dementia. Apart from peripheral neuropathy, the clinical features are non-specific, evidence for solvent-related toxicity is in most cases circumstantial and there is no
Bo Sahlberg et al.
The Science of the total environment, 444, 433-440 (2013-01-03)
There are few studies on associations between airborne microbial exposure, formaldehyde, plasticizers in dwellings and the symptoms compatible with the sick building syndrome (SBS). As a follow-up of the European Community Respiratory Health Survey (ECRHS II), indoor measurements were performed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service