Skip to Content
Merck
All Photos(6)

Key Documents

ZMS1204

Sigma-Aldrich

Anti-Mitochondria Antibody, clone 113-1 ZooMAb® Mouse Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Mitochondria Marker Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43
conjugate:
unconjugated
application:
ICC
IHC
WB
clone:
recombinant monoclonal
technique(s):
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable
citations:

biological source

mouse

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

recombinant monoclonal

description

clone 113-1
recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

observed mol wt ~60 kDa

purified by

using protein G

packaging

antibody small pack of 25

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG1κ

epitope sequence

Unknown

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry. This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 113-1 is a ZooMAb® mouse recombinant monoclonal antibody that specifically detects a 60 kDa non-glycosylated mitochondrial protein. protein..

Immunogen

A semi-purified mitochondrial preparation.

Application

Quality Control Testing

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: A 1:10,000 dilution of this antibody detected a non-glycosylated mitochondrial protein in HeLa cell lysates.

Tested Applications

Western Blotting Analysis: A 1:10,000 dilution from a representative lot detected a non-glycosylated mitochondrial protein in HepG2 cell lysate.

Immunocytochemistry Analysis: A 1:1,000 dilution from a representative lot detected a non-glycosylated mitochondrial protein in HepG2 cells.

Immunohistochemistry (Paraffin) Analysis: A 1:1,000 dilution of this antibody detected a non-glycosylated mitochondrial protein in Human heart tissue sections.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Mitochondria are important cell organelles that are responsible ATP production and biosynthesis of several intermediates. They also play a major role in cellular stress responses, autophagy, and apoptosis. They are also part of a dynamic, interconnected network that is closely integrated with other cellular compartments and their function extends beyond the boundaries of the cell. They affect the physiology of the organism by regulating communication between cells and tissues. Within a cell the mitochondria are in constant motion and can move to the areas of high energy demand and may undergo repeated rounds of fission and fusion, which is important in mixing of mitochondrial matrices and redistribution of the mitochondrial DNA. As a by-product of oxidative phosphorylation, mitochondria produce significant quantities of endogenous reactive oxygen species (ROS), which can damage cellular proteins, lipids, nuclear and mitochondrial DNA. Excessive mitochondrial ROS production can overwhelm the antioxidant defenses and can cause serious damage to mitochondria and the cell. Cumulative oxidative damage and mitochondrial DNA mutations can lead to mitochondrial abnormalities that affect the health of the organism. It has been shown that mitochondria from cancer cells are often resistant to permeabilization of their outer membranes and resist apoptosis. Clone 113-1 is an ideal antibody for staining mitochondria in human cells and can be used as a mitochondrial marker in subcellular fractions. It recognizes a 60 kDa non-glycosylated protein component of mitochondria found in human cells. It is shown to produce a spaghetti-like pattern in normal and malignant cells. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals.

Physical form

Purified recombinant mouse monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Reconstitution

300 µg/mL after reconstitution at 25 µL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service