Skip to Content
Merck
All Photos(4)

Key Documents

WH0004222M1

Sigma-Aldrich

Monoclonal Anti-MEOX1 antibody produced in mouse

clone 1A10, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-MOX1, Anti-mesenchyme homeobox 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1A10, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunofluorescence: suitable
indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MEOX1(4222)

General description

MEOX1 (mesenchyme homeobox 1) gene codes for homeobox proteins that are located on many mesodermal structures. It has an N terminal, middle, a C terminal domain, as well as a homeo domain. This gene is located on human chromosome 17q21.31.
This gene encodes a member of a subfamily of non-clustered, diverged, antennapedia-like homeobox-containing genes. The encoded protein may play a role in the molecular signaling network regulating somite development. Alternatively spliced transcript variants encoding different isoforms have been described. (provided by RefSeq)

Immunogen

MEOX1 (NP_004518.1, 165 a.a. ~ 252 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
PEGSSKARKERTAFTKEQLRELEAEFAHHNYLTRLRRYEIAVNLDLSERQVKVWFQNRRMKWKRVKGGQPISPNGQDPEDGDSTASPS

Biochem/physiol Actions

MEOX1 (mesenchyme homeobox 1) is essential in axial skeleton and limb muscle development. Mutation in MEOX1 gene results in Klippel-feil syndrome subtype.

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Homeodomain proteins Mox1 and Mox2 associate with Pax1 and Pax3 transcription factors
Stamataki D, et al.
Febs Letters, 499(3), 274-278 (2001)
Mutation in MEOX1 gene causes a recessive Klippel-Feil syndrome subtype
Bayrakli F, et al.
BMC Genetics, 14(1), 95-95 (2013)
Mutations in MEOX1, encoding mesenchyme homeobox 1, cause Klippel-Feil anomaly
Mohamed JY, et al.
American Journal of Human Genetics, 92(1), 157-161 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service