Skip to Content
Merck
All Photos(8)

Key Documents

WH0002965M1

Sigma-Aldrich

Monoclonal Anti-GTF2H1 antibody produced in mouse

clone 1F12-1B5, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-BTF2, Anti-TFIIH, Anti-general transcription factor IIH, polypeptide 1, 62kDa

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41
conjugate:
unconjugated
application:
ELISA (i)
IF
IHC (p)
WB
clone:
1F12-1B5, monoclonal
species reactivity:
human
citations:
2
technique(s):
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
indirect ELISA: suitable
indirect immunofluorescence: suitable
western blot: 1-5 μg/mL

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1F12-1B5, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
indirect ELISA: suitable
indirect immunofluorescence: suitable
western blot: 1-5 μg/mL

isotype

IgG1κ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GTF2H1(2965)

Immunogen

GTF2H1 (AAH00365, 1 a.a. ~ 548 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MATSSEEVLLIVKKVRQKKQDGALYLMAERIAWAPEGKDRFTISHMYADIKCQKISPEGKAKIQLQLVLHAGDTTNFHFSNESTAVKERDAVKDLLQQLLPKFKRKANKELEEKNRMLQEDPVLFQLYKDLVVSQVISAEEFWANRLNVNATDSSSTSNHKQDVGISAAFLADVRPQTDGCNGLRYNLTSDIIESIFRTYPAVKMKYAENVPHNMTEKEFWTRFFQSHYFHRDRLNTGSKDLFAECAKIDEKGLKTMVSLGVKNPLLDLTALEDKPLDEGYGISSVPSASNSKSIKENSNAAIIKRFNHHSAMVLAAGLRKQEAQNEQTSEPSNMDGNSGDADCFQPAVKRAKLQESIEYEDLGKNNSVKTIALNLKKSDRYYHGPTPIQSLQYATSQDIINSFQSIRQEMEAYTPKLTQVLSSSAASSTITALSPGGALMQGGTQQAINQMVPNDIQSELKHLYVAVGELLRHFWSCFPVNTPFLEEKVVKMKSNLERFQVTKLCPFQEKIRRQYLSTNLVSHIEEMLQTAYNKLHTWQSRRLMKKT

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Di Zhou et al.
Cell reports methods, 4(1), 100674-100674 (2024-01-05)
Transcription by RNA polymerase II (RNA Pol II) is crucial for cellular function, but DNA damage severely impedes this process. Thus far, transcription-blocking DNA lesions (TBLs) and their repair have been difficult to quantify in living cells. To overcome this
Arjan F Theil et al.
EMBO molecular medicine, 15(11), e17973-e17973 (2023-10-06)
The brittle hair syndrome Trichothiodystrophy (TTD) is characterized by variable clinical features, including photosensitivity, ichthyosis, growth retardation, microcephaly, intellectual disability, hypogonadism, and anaemia. TTD-associated mutations typically cause unstable mutant proteins involved in various steps of gene expression, severely reducing steady-state

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service