Skip to Content

UC9601

UltraClear Sequencing Reaction Clean-Up Kit

pkg of 1 plates, sufficient for 1 96-well plate purifications

Synonym(s):

Sequencing Clean-Up Kit, UltraClear Reaction Kit

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.52
UNSPSC Code:
41105600
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


usage

sufficient for 1 96-well plate purifications

Quality Segment

packaging

pkg of 1 plates

storage temp.

15-25°C

General description

The UltraClear Sequencing Reaction Clean-Up Plates offer a rapid and simple method for the clean-up of DNA sequencing reactions in a 96-well format. The plates use ultrafiltration membranes to separate low molecular weight contaminants, such as unincorporated dye terminators, dNTPs, and residual salts from the sequencing reaction products.

Following thermocycling, the sequencing reactions are diluted with Sequencing Solution. This mixture is then filtered through the UltraClear plate by centrifugation. The sequencing reaction products are retained on the surface of the ultrafiltration membrane while the lower molecular weight contaminants pass through the membrane and are collected as waste. The purified sequencing products are then resuspended in Sequencing Solution and are ready for injection onto capillary-based DNA analyzers.

Features and Benefits

  • Effective clean-up of ABI BigDye® chemistries
  • Simple centrifugation-based procedure
  • Convenient 96-well format

Other Notes

For additional information, please see www.sigma-aldrich.com/sequencing.

Legal Information

BigDye is a registered trademark of Applera Corporation or its subsidiaries in the US and/or certain other countries
UltraClear is a trademark of Sigma-Aldrich Co. LLC


Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library