TRUEDEXS
TrueGel3D Polymer
Modified for slow gelling (SLO-DEXTRAN), allow cell recovery
About This Item
Recommended Products
technique(s)
cell attachment: suitable
shipped in
dry ice
storage temp.
−70°C
Looking for similar products? Visit Product Comparison Guide
General description
TrueGel3D enzymatic cell recovery solution (TRUEENZ-500UL) is used to dissolve dextran polymers for easy recovery of chemically fixed or live cells for post-culture analyses including RT-PCR, western blots and other molecular biological studies.
Application
Features and Benefits
- Contains dextrose polymer functionalized with thiol reactive groups for slow gelation
- Degradable polymer
Components
Caution
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2
Storage Class Code
8A - Combustible corrosive hazardous materials
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Troubleshoot TrueGel3D™ hydrogel formation and cell viability issues with RGD-degradable polymer and CD cell-degradable crosslinker.
Troubleshoot TrueGel3D™ hydrogel formation and cell viability issues with RGD-degradable polymer and CD cell-degradable crosslinker.
Troubleshoot TrueGel3D™ hydrogel formation and cell viability issues with RGD-degradable polymer and CD cell-degradable crosslinker.
Troubleshoot TrueGel3D™ hydrogel formation and cell viability issues with RGD-degradable polymer and CD cell-degradable crosslinker.
Protocols
An advanced protocol is available for TrueGel 3D kits for 3D cell culture for peptides concentration or stiffness optimisation
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service