Skip to Content
Merck
All Photos(1)

Key Documents

T2630

Sigma-Aldrich

Glyceryl tripalmitoleate

≥98%, liquid

Synonym(s):

TG(16:1(9Z)/16:1(9Z)/16:1(9Z)), Tripalmitolein

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C51H92O6
CAS Number:
Molecular Weight:
801.27
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25
Pricing and availability is not currently available.

biological source

synthetic

Quality Level

Assay

≥98%

form

liquid

functional group

ester

lipid type

neutral glycerides

shipped in

ambient

storage temp.

−20°C

SMILES string

CCCCCC\C=C/CCCCCCCC(=O)OCC(COC(=O)CCCCCCC\C=C/CCCCCC)OC(=O)CCCCCCC\C=C/CCCCCC

InChI

1S/C51H92O6/c1-4-7-10-13-16-19-22-25-28-31-34-37-40-43-49(52)55-46-48(57-51(54)45-42-39-36-33-30-27-24-21-18-15-12-9-6-3)47-56-50(53)44-41-38-35-32-29-26-23-20-17-14-11-8-5-2/h19-24,48H,4-18,25-47H2,1-3H3/b22-19-,23-20-,24-21-

InChI key

SKGWNZXOCSYJQL-BUTYCLJRSA-N

Packaging

Sealed ampule.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Karina Salcedo-Vite et al.
Applied microbiology and biotechnology, 103(15), 6217-6229 (2019-05-31)
Acinetobacter baylyi ADP1 is a microorganism with the potential to produce storage lipids. Here, a systematic study was carried out to evaluate growth performance and accumulation of wax esters and triacylglycerols using glycerol, xylose, glucose, acetate, ethanol, and pyruvate as
Suresh K Nagumalli et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 1161, 122415-122415 (2020-11-28)
The analysis of the fatty acid profile of triglycerides has long played a central role in the evaluation and classification of edible vegetable oils. However, the range of analytical procedures available to evaluate these profiles remains limited and are typically
Alexandra Paul et al.
Biophysical journal, 116(12), 2346-2355 (2019-06-04)
Lipid droplets (LDs), present in many cell types, are highly dynamic organelles that store neutral lipids, primarily triacylglycerols (TAGs). With the discovery of new LD functions (e.g., in immune response, protein clearage, and occurrence with disease), new methods to study
Annemieke de Jong et al.
Nature immunology, 15(2), 177-185 (2013-12-24)
T cells autoreactive to the antigen-presenting molecule CD1a are common in human blood and skin, but the search for natural autoantigens has been confounded by background T cell responses to CD1 proteins and self lipids. After capturing CD1a-lipid complexes, we

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service