Skip to Content
Merck
All Photos(2)

Key Documents

SRP5091

Sigma-Aldrich

TGFβR1 (80-end), active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

AAT5, ACVRLK4, ALK-5, LDS1A, LSD2A, SKR4, TGFR-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

recombinant

expressed in baculovirus infected Sf9 cells

product line

PRECISIO® Kinase

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

2.5-3.5 nmol/min·mg

mol wt

~66 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... TGFBR1(7046)

General description

TGFβR1 or transforming growth factor, beta-receptor 1 is a member of the TGFβ receptor subfamily and is a ser/thr protein kinase that forms a heteromeric complex with type II TGF-beta receptors when bound to TGF-beta, transducing the TGF-beta signal from the cell surface to the cytoplasm. Mutations in TGFβR1gene have been associated with Marfan syndrome, Loeys-Deitz Aortic Aneurysm Syndrome, and the development of various types of tumors. TGFβR1-dependent signaling is required for angiogenesis but not for the development of hematopoietic progenitor cells and functional hematopoiesis.

Physical form

Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Krishna Kumar Singh et al.
Human mutation, 27(8), 770-777 (2006-06-27)
Marfan syndrome (MFS) is an autosomal dominant connective tissue disorder characterized by manifestations in the cardiovascular, skeletal, ocular, and other organ systems. MFS type1 (MFS1) is caused by mutations in the gene encoding fibrillin (FBN1). Recently, the transforming growth factor-beta
J Larsson et al.
The EMBO journal, 20(7), 1663-1673 (2001-04-04)
Deletion of the transforming growth factor beta1 (TGF-beta1) gene in mice has previously suggested that it regulates both hematopoiesis and angiogenesis. To define the function of TGF-beta more precisely, we inactivated the TGF-beta type I receptor (TbetaRI) gene by gene

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service