Skip to Content
Merck
All Photos(1)

Key Documents

SRP3295

Sigma-Aldrich

VAP-1 human

recombinant, expressed in CHO cells, ≥95% (SDS-PAGE), ≥95% (HPLC)

Synonym(s):

AOC3, HPAO, SSAO

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

recombinant

expressed in CHO cells

Assay

≥95% (HPLC)
≥95% (SDS-PAGE)

form

lyophilized

mol wt

80.0-90.0 kDa

packaging

pkg of 10 μg

impurities

<0.1 EU/μg endotoxin, tested

color

white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... AOC3(8639)

General description

VAP-1 is a type II membrane cell adhesion protein belonging to the copper/topaquinone oxidase family. Recombinant VAP-1 is a mixture of monomeric and disulfide linked homodimeric forms of a 737 amino acid polypeptide corresponding to amino acids 27 to 763 of the VAP-1 precursor. The gene AOC3 (amine oxidase, copper containing 3) encodes a 170kDa homodimeric sialylated glycoprotein, VAP-1 (vascular adhesion protein-1), that is abundantly expressed in high endothelial venules (HEV) of peripheral lymph nodes. It is expressed in capillaries of non-lymphatic human tissues, such as kidney and heart. It is also expressed on on adipocytes, dendritic cells and smooth muscle cells.

Biochem/physiol Actions

The protein VAP-1 (vascular adhesion protein-1) belongs to the group of semicarbazide sensitive amine oxidases (SSAO) that catalyze the formation of reactive oxygen species. It is a unique adhesion molecule that participates in inflammatory leukocyte recruitment in blood vessels. It functions in leukocyte extravasation to inflamed tissues. In the human eye, it has been associated with ocular inflammatory diseases, such as uveitis, age-related macular degeneration, and diabetic retinopathy. Its expression has been observed to be altered in several diseases, such as rheumatoid arthritis, diabetes, and atherosclerosis.

Physical form

Lyophilized from 10 mM Sodium Phosphate, pH 7.8.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/mL. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

VAP-1-mediated M2 macrophage infiltration underlies IL-1?- but not VEGF-A-induced lymph- and angiogenesis.
Nakao S
The American Journal of Pathology, 178, 1913-1921 (2011)
A J Gearing et al.
Immunology today, 14(10), 506-512 (1993-10-01)
The demonstration of soluble isoforms of adhesion molecules has added a layer of complexity of our understanding of lymphoid-endothelial cell interactions. This is especially true in the light of observations which show levels of these isoforms to be raised during
Lama Almulki et al.
Experimental eye research, 90(1), 26-32 (2009-09-19)
Recently we showed a critical role for Vascular Adhesion Protein-1 (VAP-1) in rodents during acute ocular inflammation, angiogenesis, and diabetic retinal leukostasis. However, the expression of VAP-1 in the human eye is unknown. VAP-1 localization was therefore investigated by immunohistochemistry.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service