Skip to Content
Merck

SRP0100

HDAC-1 human

recombinant, expressed in baculovirus infected insect cells, ≥50% (SDS-PAGE)

Synonym(s):

HD1, Histone deacetylase 1

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.32
UNSPSC Code:
12352200
Assay:
≥50% (SDS-PAGE)
Biological source:
human
Recombinant:
expressed in baculovirus infected insect cells
Concentration:
>0.02 mg/mL
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

human

recombinant

expressed in baculovirus infected insect cells

assay

≥50% (SDS-PAGE)

form

aqueous solution

mol wt

56 kDa

packaging

pkg of 50 μg

storage condition

avoid repeated freeze/thaw cycles

concentration

>0.02 mg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... HDAC1(3065)

General description

Human HDAC1, GenBank Accession No. NM_004964, full length with C-terminal HIS-DDDDK tag (FLAG) and C-terminal His-tag, MW = 56 kDa, expressed in baculovirus expression system.

Biochem/physiol Actions

Useful for the study of enzyme kinetics, screening inhibitors.

Physical form

Formulated in 25 mM Tris-HCl, pH 8.0, 130 mM NaCl, 0.05% Tween-20, 100 μg/ml FLAG peptide and 10% glycerol.

Preparation Note

Thaw on ice. Upon first thaw, briefly spin tube containing enzyme to recover full content of the tube. Aliquot enzyme into single use aliquots. Store remaining undiluted enzyme in aliquots at -70°C. Note: Enzyme is very sensitive to freeze/thaw cycles.

Other Notes

One unit is defined as the amount of enzyme required to deacetylate 1 pmol of substrate/min at 37°C.


Storage Class

12 - Non Combustible Liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library