Skip to Content
Merck

SML3760

TFMT

≥98% (HPLC)

Synonym(s):

(2R,3R,4S,5R)-2-(4-Amino-5-(trifluoromethyl)-7H-pyrrolo[2,3-d]pyrimidin-7-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diol, 5-Trifluoromethyl-tubercidin, 7-β-D-Ribofuranosyl-5-(trifluoromethyl)-7H-pyrrolo[2,3-d]pyrimidin-4-amine, Trifluoromethyl-tubercidin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C12H13F3N4O4
CAS Number:
Molecular Weight:
334.25
NACRES:
NA.77
UNSPSC Code:
51111800
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

Biochem/physiol Actions

SAM-binding pocket-targeting methyltransferase MTr1 inhibitor with anti-influenza efficacy in vitro and in vivo.
Trifluoromethyl-tubercidin (TFMT) is a SAM-binding pocket-targeting methyltransferase (MTase) MTr1 inhibitor that impedes the maturation of host cap RNAs and their binding with influenza viral polymerase. TFMT selectively inhibits influenza A&B (IAV & IBV), but not Hazara virus (HAZV) or soft tick bunyavirus (STBV), replication in epithelial cultures (IAV IC50 = 300 nM/human A549, 7.7 μM/murine LA-4) with little toxicity (up to 1 mM; A549 cells). TFMT suppresses IAV replication in mice in vivo (2 mg/kg intranasally on day 0 and day 1 of IAV infection) and synergizes with anti-influenza drugs baloxavir marboxil (BXM; endonuclease inhibitor) and oseltamivir (neuraminidase inhibitor).


pictograms

Skull and crossbones

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 2 Oral

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library