Skip to Content
Merck
All Photos(1)

Key Documents

SML3411

Sigma-Aldrich

LY2828360

≥98% (HPLC)

Synonym(s):

8-(2-Chlorophenyl)-2-methyl-6-(4-methyl-1-piperazinyl)-9-(tetrahydro-2H-pyran-4-yl)-9H-purine, LY 2828360

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H27ClN6O
CAS Number:
Molecular Weight:
426.94
MDL number:
UNSPSC Code:
41121800
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (Warmed)

storage temp.

-10 to -25°C

SMILES string

Clc1c(cccc1)c2[n](c4nc(nc(c4n2)N5CCN(CC5)C)C)C3CCOCC3

InChI

1S/C22H27ClN6O/c1-15-24-21(28-11-9-27(2)10-12-28)19-22(25-15)29(16-7-13-30-14-8-16)20(26-19)17-5-3-4-6-18(17)23/h3-6,16H,7-14H2,1-2H3

InChI key

UCMNDPDJRSEZPL-UHFFFAOYSA-N

Biochem/physiol Actions

LY2828360 is a potent and selective slowly acting G protein-biased CB2 cannabinoid agonist that inhibits cAMP accumulation and activates ERK1/2 signaling while failing to recruit arrestin. LY2828360 suppress chemotherapy-induced neuropathic pain created by paclitaxel without producing tolerance in mice.
Potent and selective slowly acting G protein-biased CB2 cannabinoid agonist

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service