Skip to Content
Merck

SML1879

A-395N

≥98% (HPLC)

Synonym(s):

(3S,4R)-1-Benzyl-N,N-dimethyl-4-(4-(4-(methylsulfonyl)piperazin-1-yl)phenyl)pyrrolidin-3-amine, (3S,4R)-{1-Benzyl-4-[4-(4-methanesulfonyl-piperazin-1-yl)-phenyl]-pyrrolidin-3-yl}-dimethyl-amine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C24H34N4O2S
CAS Number:
Molecular Weight:
442.62
NACRES:
NA.77
UNSPSC Code:
12352204
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

optical activity

[α]/D +1.0 to +3.0°, c = 1 in chloroform-d

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

2-8°C

Biochem/physiol Actions

A-395N is a control probe for A-395, which is a chemical probe for polycomb protein EED. A-395 is a potent and selective chemical probe for the polycomb protein EED (embryonic ectoderm development), an essential component of Polycomb repressive complex 2 (PRC2), involved in transcriptional repression through methylation of histone H3K27. A-395N is structurally similar, but exhibits no activity in the biochemical and cellular assays, so is an ideal control compound. For characterization details of the active probe, A-395, please visit the A-395 probe summary on the Structural Genomics Consortium (SGC) website.

A-395, the active enantiomer, is available from Sigma. To learn more about and purchase A-395, click here.

To learn about other SGC chemical probes for epigenetic targets, visit sigma.com/sgc
A-395N is a control probe for A-395, which is a chemical probe for polycomb protein EED..

Features and Benefits

A-395N is a negative control for A-395, an epigenetic chemical probe available through a partnership with the Structural Genomics Consortium (SGC). To learn more and view other SGC epigenetic probes, visit sigma.com/SGC.


pictograms

Skull and crossbones

signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library