Skip to Content
Merck

SML1604

NDB

≥98% (HPLC)

Synonym(s):

Benzamide, 2,6-dichloro-4-(dimethylamino)-N-[3-(1,1-dimethylethyl)-4-hydroxyphenyl]-N-(phenylmethyl)-, N-Benzyl-N-(3-tert-butyl-4-hydroxy-phenyl)-2,6-dichloro-4-dimethylamino-benzamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C26H28Cl2N2O2
CAS Number:
Molecular Weight:
471.42
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

2-8°C

SMILES string

CN(C)C1=CC(Cl)=C(C(N(CC2=CC=CC=C2)C3=CC(C(C)(C)C)=C(O)C=C3)=O)C(Cl)=C1

InChI

1S/C26H28Cl2N2O2/c1-26(2,3)20-13-18(11-12-23(20)31)30(16-17-9-7-6-8-10-17)25(32)24-21(27)14-19(29(4)5)15-22(24)28/h6-15,31H,16H2,1-5H3

InChI key

IDACWMHIKWNAEO-UHFFFAOYSA-N

Biochem/physiol Actions

NDB is a selective antagonist of human Farnesoid X receptor α (FXRα) that effectively modulates FXRα down-stream genes.
NDB is a selective antagonist of human Farnesoid X receptor α.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library