Skip to Content
Merck

SML1464

SMBA1

≥98% (HPLC)

Synonym(s):

2-[(2-Nitro-9H-fluoren-9-ylidene)methyl]-phenol, 2-[(2-Nitro-9H-fluorene-9-ylidene)methyl]phenol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C20H13NO3
CAS Number:
Molecular Weight:
315.32
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

−20°C

SMILES string

OC(C=CC=C1)=C1/C=C2C(C=CC=C3)=C3C4=C\2C=C([N+]([O-])=O)C=C4

InChI

1S/C20H13NO3/c22-20-8-4-1-5-13(20)11-18-16-7-3-2-6-15(16)17-10-9-14(21(23)24)12-19(17)18/h1-12,22H/b18-11+

InChI key

PEYKRJBXACUOOB-WOJGMQOQSA-N

Biochem/physiol Actions

SMBA1 is a potent and selective Bax agonist.
SMBA1 is an anti-tumor agent. It exhibits anti-proliferative effects against glioblastoma (GBM) cells.
Small-molecule Bax agonists 1 (SMBA1) is a potent and selective Bax agonist that binds to Bax, induces conformational change which blocks Bax S184 phosphorylation. SMBA1 induces apoptosis in cancer cells and suppresses lung tumor growth in vivo without significant normal tissue toxicity. SMBA1 does not bind to other B-cell lymphoma 2 (Bcl-2) family members.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library