Skip to Content
Merck

SML1064

IMM-01

≥98% (HPLC)

Synonym(s):

2-[(2,4-Dihydroxyphenyl)methylene]-N-(1,1-dimethylethyl)-hydrazinecarbothioamide, Intramimic-01, N1-(tert-Butyl)-2-(2,4-dihydroxybenzylidene)hydrazine-1-carbothioamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C12H17N3O2S
CAS Number:
Molecular Weight:
267.35
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to light brown

solubility

DMSO: 10 mg/mL, clear

storage temp.

2-8°C

SMILES string

S=C(N\N=C\c1c(cc(cc1)O)O)NC(C)(C)C

InChI

1S/C12H17N3O2S/c1-12(2,3)14-11(18)15-13-7-8-4-5-9(16)6-10(8)17/h4-7,16-17H,1-3H3,(H2,14,15,18)/b13-7+

InChI key

LTFUAYRGVLQXKC-NTUHNPAUSA-N

Application

IMM-01 has been used as a formin activator to study its effects on motor neurons (MNs).

Biochem/physiol Actions

IMM-01 binds and activates formin thereby leading to autoinhinition.
IMM-01 is an agonist of the mammalian Diaphanous (mDia)-related formins that triggers actin assembly and microtubule stabilization, serum response factor-mediated gene expression, cell-cycle arrest, and apoptosis. IMM-01 inhibits DID–DAD binding, which in turn activates mDia family of formins in cells. IMM-01 slows tumor growth in a mouse xenograft model of colon cancer.
IMM-01 is an agonist of the mammalian Diaphanous (mDia)-related formins that triggers actin assembly.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library