Skip to Content
Merck
All Photos(1)

Key Documents

SML0884

Sigma-Aldrich

bpV(HOpic)

≥90% V basis

Synonym(s):

Bisperoxovanadium(HOpic), Dipotassium bisperoxo (5-hydroxypyridine-2-carboxyl) oxovanadate (V), K2[VO(O2)2C6H4NO3]

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
K2VOO2O2C6H4NO3
CAS Number:
Molecular Weight:
347.24
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥90% V basis

form

powder

storage condition

desiccated
protect from light

color

faintly yellow to dark yellow

solubility

H2O: 20 mg/mL, clear

storage temp.

−20°C

SMILES string

[V]32(OO3)(OO2)([NH]1=CC(=O)CC=C1C(=O)[O-])[O-].[K+].[K+].O.O.O.O

InChI

1S/C6H5NO3.2K.2O2.4H2O.O.V/c8-4-1-2-5(6(9)10)7-3-4;;;2*1-2;;;;;;/h2-3H,1H2,(H,9,10);;;;;4*1H2;;/q;2*+1;2*-2;;;;;-1;+4/p-1

InChI key

HSCJTESZNNRNPK-UHFFFAOYSA-M

Application

bpV(HOpic) has been used in pharmacological perturbation of cell migration related signaling pathway.

Biochem/physiol Actions

bpV(OHpic) is a bisperoxovanadium inhibitor of protein phosphotyrosine phosphatases with selectivity for PTEN, phosphatase and tensin homolog, a tumour suppressor phosphatase involved in cell cycle regulation. IC50 values for bpV(HOpic) are 14 nM for PTEN compared to 4.9 μM for PTPβ and 25.3 μM for PTP-1β. bpV(HOpic) has been shown to enhance PI3K/Akt signaling that could prevent myocardium from ischemia-reperfusion injury.

Other Notes

Soluble in DMSO or water. NOTE: The product is not stable in water. Decomposition starts immediately after solubilization. Rate of decomposition is slower in dry DMSO.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Directed migration of cancer cells guided by the graded texture of the underlying matrix.
Park J, et al.
Nature Materials, 15(7), 792-792 (2016)
Thomas Theis et al.
Molecular neurobiology, 55(10), 8203-8215 (2018-03-10)
Glycosylation fine-tunes signal transduction of adhesion molecules during neural development and supports synaptic plasticity and repair after injury in the adult nervous system. One abundantly expressed neural glycan is LewisX (LeX). Although it is known that its expression starts at

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service