Skip to Content
Merck

SML0690

L-161,982

≥98% (HPLC)

Synonym(s):

N-[[4′-[[3-Butyl-1,5-dihydro-5-oxo-1-[2-(trifluoromethyl)phenyl]-4H-1,2,4-triazol-4-yl]methyl][1,1"-biphenyl]-2-yl]sulfonyl]-3-methyl-2-thiophenecarboxamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C32H29F3N4O4S2
CAS Number:
Molecular Weight:
654.72
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 15 mg/mL, clear

storage temp.

2-8°C

SMILES string

FC(F)(F)c1c(cccc1)[n]2nc([n]([c]2=O)Cc3ccc(cc3)c4c(cccc4)[S](=O)(=O)NC(=O)c5[s]ccc5C)CCCC

InChI

1S/C32H29F3N4O4S2/c1-3-4-13-28-36-39(26-11-7-6-10-25(26)32(33,34)35)31(41)38(28)20-22-14-16-23(17-15-22)24-9-5-8-12-27(24)45(42,43)37-30(40)29-21(2)18-19-44-29/h5-12,14-19H,3-4,13,20H2,1-2H3,(H,37,40)

InChI key

MMDNKTXNUZFVKD-UHFFFAOYSA-N

Biochem/physiol Actions

L-161,982 is a potent EP4 receptor antagonist that is selective over all other members of the prostanoid receptor family (Ki values are 0.024, 0.71, 1.90, 5.10, 5.63, 6.74, 19 and 23 μM for human EP4, TP, EP3, DP, FP, IP, EP1 and EP2 receptors respectively.
L-161,982 is a potent EP4 receptor antagonist.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library