Skip to Content
Merck
All Photos(1)

Key Documents

SML0635

Sigma-Aldrich

AZD6765 dihydrochloride

≥97% (HPLC)

Synonym(s):

(αS)-α-Phenyl-2-pyridineethanamine dihydrochloride, AR-R 15896 dihydrochloride, FPL 15896 dihydrochloride, Lanicemine dihydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H14N2 · 2HCl
CAS Number:
Molecular Weight:
271.19
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥97% (HPLC)

form

powder

optical activity

[α]/D +77 to +90°, c = 1 in methanol

storage condition

desiccated

color

white to beige

solubility

H2O: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

[Cl-].[Cl-].[N+H3][C@@H](Cc2[n+H]cccc2)c1ccccc1

InChI

1S/C13H14N2.2ClH/c14-13(11-6-2-1-3-7-11)10-12-8-4-5-9-15-12;;/h1-9,13H,10,14H2;2*1H/t13-;;/m0../s1

InChI key

KHJHFYAGQZYCLC-GXKRWWSZSA-N

Application

AZD6765 dihydrochloride has been used as an antidepressant to improve behavior and metabolic measures in chronic unpredictable mild stress (CUMS).

Biochem/physiol Actions

AZD6765 is a noncompetitive N-methyl-D-aspartate (NMDA) receptor antagonist and fast-acting antidepressant. In a recent study AZD6765 relieved depression within 80 minutes, unlike most antidepressants acting on serotonin or dopamine systems which can take several weeks to work. The effects lasted from about an hour up to 2 weeks. AZD6765 has an IC50 value of 1.3 μM. Unlike more potent drugs such as ketamine, which also has rapid antidepressant activity, AZD6765 has no dissociative side-effects such as hallucinations. Earlier studies inidcated the compound has neuroprotective and anticonvulsant effects.

Features and Benefits

This compound is featured on the Glutamate Receptors (Ion Channel Family) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pravin Kumar Mishra et al.
Neurochemistry international, 137, 104750-104750 (2020-05-04)
Major depressive disorder is the leading cause of disability and suicidality worldwide. Here, we evaluated neural metabolic activity in prefrontal cortex (PFC) in C57BL6 mice undergoing a chronic unpredictable mild stress (CUMS) for three weeks to induce depression. Further, the

Articles

We offers many products related to ion channel family glutamate receptors for your research needs.

We offers many products related to ion channel family glutamate receptors for your research needs.

We offers many products related to ion channel family glutamate receptors for your research needs.

We offers many products related to ion channel family glutamate receptors for your research needs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service