Skip to Content
Merck
All Photos(1)

Documents

SMB00312

Sigma-Aldrich

3-N-Butylphthalide

≥98% (HPLC)

Synonym(s):

3-N-Butylphthalide, 3-Butylphthalide, 3-N-butyl-phthalide, butylphthalide, n-Butylphthalide, 3-Butylisobenzofuran-1(3H)-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H14O2
CAS Number:
Molecular Weight:
190.24
EC Number:
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.25

Quality Level

Assay

≥98% (HPLC)

form

liquid

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

O=C1C2=CC=CC=C2C(CCCC)O1

InChI

1S/C12H14O2/c1-2-3-8-11-9-6-4-5-7-10(9)12(13)14-11/h4-7,11H,2-3,8H2,1H3

InChI key

HJXMNVQARNZTEE-UHFFFAOYSA-N

General description

3-N-Butylphthalide (NBP) is a compound found in Chinese celery seed extracts that can improve cognitive functions and may decrease Amyloid-beta levels in Alzheimer′s disease. NBP has antioxidant activities and may protect against oxidative/nitrosative stress, mitochondrial impairment and apoptosis.

Preparation Note

Centrifuge vial before use to aid in recovery of material.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service