Skip to Content
Merck
All Photos(1)

Key Documents

SCP0147

Sigma-Aldrich

FR-1, cyclic

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C35H57N13O16S2
Molecular Weight:
980.03
UNSPSC Code:
12352202
NACRES:
NA.32

Assay

≥95% (HPLC)

form

lyophilized

composition

Peptide Content, ≥78%

storage condition

protect from light

storage temp.

−20°C

Amino Acid Sequence

Cys-Arg-Gly-Asp-Ser-Pro-Ala-Ser-Ser-Cys

Application

FR-1 is a cyclic RGDS peptide that acts via binding to FR-1 receptor, a 57 kDa Arg-Gly-Asp-Ser (RGDS)-binding protein involved in early embryogenesis.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H Katow et al.
Development, growth & differentiation, 43(5), 601-610 (2001-09-29)
Immunoblotting using polyclonal antibodies (pAb) raised against an FR-1 receptor (FR-1R), a 57 kDa Arg-Gly-Asp-Ser (RGDS)-binding protein, of the sand dollar Clypeaster japonicus showed that the pAb monospecifically bound to the protein. FR-1R was present in purified plasma membrane, suggesting
H Katow et al.
Development, growth & differentiation, 39(2), 207-219 (1997-04-01)
A fibronectin-related synthetic cyclic H-Cys-Arg-Gly-Asp-Ser-Pro-Ala-Ser-Ser-Cys-OH (RGDSPASS) peptide (FR-1) binding site in the embryo of the sand dollar Clypeaster japonicus was specified using dansyl-labeled FR-1 (Dns-FR-1) and horseradish peroxidase-labeled FR-1, and an FR-1 receptor was isolated using FR-1-affinity column chromatography. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service