Skip to Content
Merck
All Photos(3)

Documents

SAB1405394

Sigma-Aldrich

Monoclonal Anti-ANGPTL5 antibody produced in mouse

clone 1B2, ascites fluid

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

1B2, monoclonal

mol wt

antigen ~68.79 kDa

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1:500-1:1000

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

Immunogen

ANGPTL5 (AAH49170.1, 1 a.a. ~ 388 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MMSPSQASLLFLNVCIFICGEAVQGNCVHHSTDSSVVNIVEDGSNAKDESKSNDTVCKEDCEESCDVKTKITREEKHFMCRNLQNSIVSYTRSTKKLLRNMMDEQQASLDYLSNQVNELMNRVLLLTTEVFRKQLDPFPHRPVQSHGLDCTDIKDTIGSVTKTPSGLYIIHPEGSSYPFEVMCDMDYRGGGWTVIQKRIDGIIDFQRLWCDYLDGFGDLLGEFWLGLKKIFYIVNQKNTSFMLYVALESEDDTLAYASYDNFWLEDETRFFKMHLGRYSGNAGDAFRGLKREDNQNAKPFSTSDVDNDGCRPACLVNGQSVKSCSHLHNKTGWWFNECGLANLNGIHHFSGKLLATGIQWGTWTKNNSPVKIKSVSMKIRRMYNPYFK

Physical form

Clear solution

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service