Skip to Content

PLA0205

Rabbit anti-CREB Antibody, Affinity Purified

Powered by Bethyl Laboratories, Inc.

Synonym(s):

5′-monophosphate response element-binding protein CREB, CREB, CREB-1, active transcription factor CREB, cAMP-response element-binding protein-1, cAMP-responsive element-binding protein 1, cyclic adenosine 3′, transactivator protein

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.41
UNSPSC Code:
12352203
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IHC, IP, WB
Citations:
1
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, human

technique(s)

immunohistochemistry: 1:100- 1:1,000, immunoprecipitation (IP): 2-5 μg/mg, western blot: 1:2,000- 1:10,000

accession no.

NP_604391.1

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

rabbit ... CREB(1385)

Immunogen

The epitope recognized by PLA0205 maps to a region between residue 250 and 300 of human cAMP responsive element binding protein 1 using the numbering given in entry NP_604391.1 (GeneID 1385).

Physical form

Tris-buffered Saline containing 0.1% BSA containing 0.09% Sodium Azide

Other Notes

CREB (cyclic AMP response element-binding protein) is a ubiquitously expressed transcription factor that binds cAMP response elements to regulate gene transcription for various cellular responses such as glucose homeostasis, growth-factor dependent survival, and proliferation. CREB is also implicated to play a role in memory and mood.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library