Skip to Content
Merck
All Photos(1)

Key Documents

MTOX1020

Sigma-Aldrich

HepaRG Cells MRP2 (-/-)

human female liver (hepatocarcinoma and hepatitis C tumor)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.81
biological source:
human female liver (hepatocarcinoma and hepatitis C tumor)

Product Name

HepaRG Cells MRP2 (-/-), one vial

biological source

human female liver (hepatocarcinoma and hepatitis C tumor)

Quality Level

form

liquid

OMIM accession no.

storage temp.

−196°C

Gene Information

human ... ABCC2(1244)

General description

HepaRG is a human hepatoma cell line. The cells possess a pseudodiploid karyotype and have been characterized as an oval ductular bipotent hepatic cell line as they have the ability to differentiate into both biliary and hepatocyte lineages in the presence of DMSO. HepaRG MRP2 knockout cells express the major xenobiotic sensors (PXR, CAR and AhR), drug transporters, phase I and II drug metabolizing enzymes as well as key hepatic transcription factors involved in stress response pathways.

Application

See technical bulletin for detailed protocols

Features and Benefits

HepaRG cells are the most metabolically active human hepatocyte cell line developed to date. These the cells are suitable for a wide variety of studies for drug metabolism, CYP induction, metabolism-mediated toxicity, transporter, and hepatotoxicity. - Sigma′s HepaRG MRP2 Knockout (KO) allows investigations of drug-transporter interactions involving MRP2 in the liver.
Zinc finger nucleases (ZFN) mediated Knockout of ABCC2 (MRP2) gene.
  • The frame-shift mutation of ABCC2 gene was confirmed by fragment length analysis and DNA sequencing.
  • Loss of functionality was confirmed by loss of transport of selective substrates in sandwich culture assay.

Quality

Tested for Mycoplasma, sterility, post-freeze viability, short terminal repeat (STR) analysis for cell line identification, cytochrome oxidase I (COI) analysis for cell line species confirmation.

Legal Information

Exhibit 1: ADME/Tox cell lines license
Exhibit 2: HepaRG limited use license
HepaRG is a trademark of BioPredic International company

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Oral drug delivery involves dissolution in the small intestine and absorption across the enterocyte barrier into the portal vein followed by subsequent delivery through the liver into the systemic circulation.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service