Merck
MTCA8813
MTC Bio Blu-Lite UHC Autoradiography Flim
W × L 5 in. × 7 in.
Synonym(s):
blotting film, western imaging film, MTC Bio A8813, imaging film, western blotting film
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NB.15
UNSPSC Code:
41105341
Manufacturer/tradename:
MTC Bio A8813
W × l:
5 in. × 7 in.
Packaging:
pack of 100 ea
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
packaging
pack of 100 ea
manufacturer/tradename
MTC Bio A8813
W × L
5 in. × 7 in.
General description
Blu-Lite UHC Autoradiography film, 5x7in, 100 sheets/box
Features and Benefits
  • Medium/dark blue base
  • Ideal for chemiluminescence
  • Ultra-high contrast
  • Ideal for 14C, 32P, 33P, 125I, and 35S
Legal Information
Blu-Lite is a trademark of MTC Bio
MTC is a trademark of MTC Bio
Still not finding the right product?
Choose from one of the most recent versions:
Certificates of Analysis (COA)