Merck
MTCA8805
MTC Bio BluBlot HS Autoradiography Flim
W × L 8 in. × 10 in.
Synonym(s):
blotting film, western imaging film, MTC Bio A8805, imaging film, western blotting film
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NB.15
UNSPSC Code:
41105341
Manufacturer/tradename:
MTC Bio A8805
W × l:
8 in. × 10 in.
Packaging:
pack of 100 ea
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
packaging
pack of 100 ea
manufacturer/tradename
MTC Bio A8805
W × L
8 in. × 10 in.
General description
BluBlot HS Autoradiography film, 8x10in, 100 sheets/box
Features and Benefits
  • High Contrast
  • Reduced Background
  • Publication Quality
  • Suitable for 14C, 32P, 33P, 125I, and 35S
  • Double Emulsion
Legal Information
BluBlot is a trademark of MTC Bio
MTC is a trademark of MTC Bio
Still not finding the right product?
Choose from one of the most recent versions:
Certificates of Analysis (COA)