Skip to Content
Merck

Skip To

HS2860E

TrueNorth® Cardboard Cryovial Box Cell Divider

Holds 100 x 1.5-2.0 mL tubes, pk of 12

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NB.22
UNSPSC Code:
41123304
Manufacturer/tradename:
Heathrow Scientific HS2860E
Feature:
Holds 100 x 1.5-2.0 mL tubes
Material:
cardboard
Packaging:
pk of 12
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

cardboard

feature

Holds 100 x 1.5-2.0 mL tubes

packaging

pk of 12

manufacturer/tradename

Heathrow Scientific HS2860E

technique(s)

cryopreservation: suitable

well diam.

11.5 mm

Application

TrueNorth® Cardboard Cryovial Box Cell Divider may be suitable for the storage of small samples and tubes.

Features and Benefits

  • for storage of small samples and tubes
  • store in LN2 or in mechanical freezers
  • colors allow for easy identification and sorting purposes
  • bottom vent holes to allow quicker circulation and removal of cold air, vapor, or LN
  • all box footprints are 13.3 x 13.3 cm, select from two different heights

  • Store in mechanical freezers or vapor phase LN2, -196ºC.
  • Colors facilitate identifying and sorting purposes with ease.
  • Bottom vent holes aid in faster circulation and elimination of cold air, vapor, or LN2
  • All box footprints are 13.3 * 13.3 cm, select from two different heights
  • Use tubes capable of being stored at the required temperature of the user.
  • Not recommended for autoclaving.

Legal Information

TrueNorth is a registered trademark of Heathrow Scientific LLC

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
HS2860FHS2860GHS2860H
description

Holds 100 x 1.5-2.0 mL tubes, pk of 12

description

Holds 64 x 1.5-2.0 mL tubes, pk of 12

description

Holds 49 x 1.5-2.0 mL tubes, pk of 12

description

Holds 81 x 1.5-2.0 mL tubes, pk of 12

manufacturer/tradename

Heathrow Scientific HS2860E

manufacturer/tradename

Heathrow Scientific HS2860F

manufacturer/tradename

Heathrow Scientific HS2860G

manufacturer/tradename

Heathrow Scientific HS2860H

feature

Holds 100 x 1.5-2.0 mL tubes

feature

Holds 64 x 1.5-2.0 mL tubes

feature

Holds 49 x 1.5-2.0 mL tubes

feature

Holds 81 x 1.5-2.0 mL tubes

material

cardboard

material

cardboard

material

cardboard

material

cardboard

packaging

pk of 12

packaging

pk of 12

packaging

pk of 12

packaging

pk of 12

well diam.

11.5 mm

well diam.

15 mm

well diam.

17.5 mm

well diam.

12 mm

technique(s)

cryopreservation: suitable

technique(s)

cryopreservation: suitable

technique(s)

cryopreservation: suitable

technique(s)

cryopreservation: suitable


Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library