Skip to Content
Merck
All Photos(1)

Key Documents

HS1421C

Weigh Boat

Large, square blue polystyrene, pk of 500 ea

Synonym(s):

weigh boat, weigh dish, weighing dish, weighing pan

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121812
NACRES:
NB.35

material

square blue polystyrene

size

large

packaging

pk of 500 ea

manufacturer/tradename

Heathrow Scientific HS1421C

L × W × H

(140 mm) × 140 mm × 22 mm

color

blue

Looking for similar products? Visit Product Comparison Guide

General description

Flexible polystyrene boats feature smooth surfaces to provide accurate pour-outs with minimal sample loss.

Features and Benefits

  • biologically inert and resistant to dilute and weak acids, aqueous solutions, alcohols, and bases.
  • temperature range -10 to 70C
  • disposable
  • shallow with a wide, flat bottom to resist tipping
  • rounded corners simplify transfer


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service