Skip to Content
Merck
All Photos(6)

Key Documents

HPA014658

Sigma-Aldrich

Anti-ARHGEF11 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-PDZ-RhoGEF, Anti-Rho guanine nucleotide exchange factor 11

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
independent
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

FIICTSKLGPPQIYELVALTSSDKNTWMELLEEAVRNATRHPGAAPMPVHPPPPGPREPAQQGPTPSRVELDDSDVFHGEPEPEELPGGTGSQQRVQGKHQVLLEDPEQEGSAEEEELGVLPCPSTSLDGENRGIR

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ARHGEF11(9826)

General description

ARHGEF11 (Rho guanine nucleotide exchange factor (GEF) 11) belongs to the family of Rho guanine nucleotide exchange factors (GEFs). It shares homology to ARHGEF1 and ARHGEF12, and regulates G-protein signaling. This gene maps to human chromosome 1q21, and is expressed in a wide range of tissues such as, adipose tissue, liver, pancreas and muscle. This protein contains a PDZ (PSD-95, Disc-large, ZO1) domain and a G protein signaling (RGS) domain.

Immunogen

Rho guanine nucleotide exchange factor 11 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

ARHGEF11 (Rho guanine nucleotide exchange factor (GEF) 11) induces Rho-GTPases, which modulate G protein signaling. It is involved in the regulation of lipid metabolism, insulin secretion and signaling. It also plays a part in axonal guidance. ARHGEF11 contains an actin-binding domain, and therefore, may play a role in determining the structure of actin cytoskeleton. R1467H variant of this gene might be responsible for increased susceptibility to type 2 diabetes mellitus in Chinese and German Caucasian population. It also interacts with RhoA, myosin II, and actomyosin, to establish cell polarity.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70693

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alejandro Castillo-Kauil et al.
The Journal of biological chemistry, 295(50), 16920-16928 (2020-10-08)
Gα proteins promote dynamic adjustments of cell shape directed by actin-cytoskeleton reorganization via their respective RhoGEF effectors. For example, Gα13 binding to the RGS-homology (RH) domains of several RH-RhoGEFs allosterically activates these proteins, causing them to expose their catalytic Dbl-homology
Rodolfo Daniel Cervantes-Villagrana et al.
International journal of molecular sciences, 24(21) (2023-11-14)
Oncogenic Gαq causes uveal melanoma via non-canonical signaling pathways. This constitutively active mutant GTPase is also found in cutaneous melanoma, lung adenocarcinoma, and seminoma, as well as in benign vascular tumors, such as congenital hemangiomas. We recently described that PDZ-RhoGEF
Kit Wong et al.
The Journal of cell biology, 179(6), 1141-1148 (2007-12-19)
Chemoattractants such as formyl-Met-Leu-Phe (fMLP) induce neutrophils to polarize by triggering divergent pathways that promote formation of a protrusive front and contracting back and sides. RhoA, a Rho GTPase, stimulates assembly of actomyosin contractile complexes at the sides and back.
Yvonne Böttcher et al.
Journal of human genetics, 53(4), 365-367 (2008-01-31)
The human rho guanine nucleotide exchange factor 11 (ARHGEF11) functions as an activator of rho GTPases and is supposed to influence insulin signalling. We investigated the effects of the previously reported R1467H variant in individual's susceptibility to type 2 diabetes
Jing Liu et al.
Molecular biology reports, 38(4), 2499-2505 (2011-01-07)
The human Rho guanine nucleotide exchange factor 11 (ARHGEF11), located on chromosome 1q21, is an activator of Rho GTPases involved in G protein signaling pathway known to regulate insulin secretion and action. The aim of our study was to evaluate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service