Skip to Content
Merck
All Photos(2)

Key Documents

H6759

Sigma-Aldrich

L-Histidine β-naphthylamide

≥98% (TLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H16N4O
CAS Number:
Molecular Weight:
280.32
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

product name

L-Histidine β-naphthylamide, powder

Assay

≥98% (TLC)

form

powder

color

white to faint yellow

storage temp.

−20°C

SMILES string

N[C@@H](Cc1cnc[nH]1)C(=O)Nc2ccc3ccccc3c2

InChI

1S/C16H16N4O/c17-15(8-14-9-18-10-19-14)16(21)20-13-6-5-11-3-1-2-4-12(11)7-13/h1-7,9-10,15H,8,17H2,(H,18,19)(H,20,21)/t15-/m0/s1

InChI key

DKDILZBBFKZMRO-HNNXBMFYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

L-Histidine β-naphthylamide is an analogue of L-histidine used to study structural roles of L-histidine in functions such as the activation of HutP protein transcription. L-Histidine β-naphthylamide may also be used to study the specificity and kinetics of various proteases and peptidases.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Allosteric activation of HutP protein, that regulates transcription of hut operon in Bacillus subtilis, mediated by various analogs of histidine.
Kumarevel T, Mizuno H, Kumar PK.
Nucleic Acids Research, 3, 199-200 (2003)
X Iturrioz et al.
Biochemistry, 39(11), 3061-3068 (2000-03-15)
Aminopeptidase A (EC 3.4.11.7, APA) is a 130 kDa membrane-bound protease that contains the HEXXH consensus sequence found in the zinc metalloprotease family, the zincins. In addition to the catalytic zinc atom, APA contains a Ca2+ ion that increases its
R A Field et al.
The Biochemical journal, 274 ( Pt 3), 885-889 (1991-03-15)
This present study reports the ability of a range of derivatives of L-histidine, histamine and imidazole to act as inhibitors of sweet-almond beta-glucosidase, yeast alpha-glucosidase and Escherichia coli beta-galactosidase. The addition of a hydrophobic group to the basic imidazole nucleus
Endang Setyorini et al.
Journal of basic microbiology, 46(4), 294-304 (2006-07-19)
A halotolerant strain FP-133, able to grow at concentrations of 0-12.5% (w/v) NaCl, was isolated from a fish paste and identified as Bacillus subtilis . B. subtilis strain FP-133 produced an intracellular protease which showed catalytic activity under saline conditions.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service