Skip to Content

GE29321908

Superdex® 75 Increase HiScale®

column thread × L (26/40) (40 cm), Cytiva 29321908

Synonym(s):

SEC column, chromatography column, prepacked column, size exclusion chromatography column

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NB.21
UNSPSC Code:
41115709
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


feature

Samples volume up to 8.5 mL

manufacturer/tradename

Cytiva 29321908

parameter

2 Mpa max. pressure

bed volume × D × L

(212 mL) (26 mm) (400 mm)

column thread × L

(26/40) (40 cm)

General description

HiScale columns, packed with new-generation SEC resins, provide the highest resolution among preparative SEC columns from Cytiva.

If you like our new-generation Superdex Increase and Superose Increase size exclusion chromatography (SEC) columns, but need to purify larger volumes of up to 8.5 mL, we can pack the resin for you in HiScale columns.

HiScale columns are packed on demand for you with Superdex 200 Increase, Superdex 75 Increase, and Superose 6 Increase SEC (gel filtration) resins. The columns are available in two sizes: 16 × 400 mm and 26 × 400 mm. These two column sizes enable purification of sample volumes up to 3.2 and 8.5 mL, respectively.

Features and Benefits

  • High-resolution SEC columns—small particle size and narrow particle size distribution of the packed resin ensures highly pure protein.
  • HiScale columns packed with SEC resins on demand, please request a quote.

Legal Information

HiScale is a registered trademark of Cytiva
Superdex is a registered trademark of Cytiva


Still not finding the right product?

Explore all of our products under Superdex® 75 Increase HiScale®


Pictograms

Flame

Signal Word

Warning

Hazard Codes

Hazard Classifications

Flam. Liq. 3

Storage Class

3 - Flammable liquids

WGK

WGK 1

Flash Point (°F)

96.8 °F

Flash Point (°C)

36 °C



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library