GBL622102
Grace Bio-Labs CoverWell™ perfusion chambers
1 chamber, port diam. 1.5 mm, chamber W × L × thickness 19 mm × 32 mm × 1.7 mm
Synonym(s):
leak proof enclosure for slides, microscope stick on multiwell chamber, reusable press to seal multiwell chamber, water tight multiple well chamber
About This Item
Recommended Products
material
clear polycarbonate backing
red silicon wells
feature
wells 1
packaging
pack of 20 ea
manufacturer/tradename
Grace Bio-Labs 622102
chamber W × L × thickness
19 mm × 32 mm × 1.7 mm
port diam.
1.5 mm
wells
1
Related Categories
General description
CoverWell perfusion chambers form removable, water-tight multiwell enclosures by pressing to coverglass or microscope slides. Chambers are made of medical-grade silicone bonded to clear, UV-transparent plastic, and are removable for permanent mounting. The thin polycarbonate cover displays excellent transparency. The perfusion chambers are ideal for live cell imaging on upright or inverted microscopes at high magnification.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)