Skip to Content
Merck

E5161

Enniatin A1

From Gnomonia errabunda, ≥95% (HPLC), mycotocin

Synonym(s):

2-(N-Methyl-L-valine) Enniatin A, Cyclo[(2R)-2-hydroxy-3-methylbutanoyl-N-methyl-L-isoleucyl-(2R)-2-hydroxy-3-methylbutanoyl-N-methyl-L-isoleucyl-(2R)-2-hydroxy-3-methylbutanoyl-N-methyl-L-valyl]

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C35H61N3O9
CAS Number:
Molecular Weight:
667.87
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥95% (HPLC)
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Enniatin A1, from Gnomonia errabunda, ≥95% (HPLC)

biological source

Gnomonia errabunda

Quality Segment

assay

≥95% (HPLC)

solubility

DMSO: 10 mg/mL, ethanol: 10 mg/mL, methanol: 10 mg/mL

storage temp.

−20°C

SMILES string

N1([C@H](C(=O)O[C@@H](C(=O)N([C@H](C(=O)O[C@@H](C(=O)N([C@H](C(=O)O[C@@H](C1=O)C(C)C)[C@H](CC)C)C)C(C)C)C(C)C)C)C(C)C)[C@H](CC)C)C

InChI

1S/C35H61N3O9/c1-16-22(11)25-34(43)46-27(19(5)6)30(39)36(13)24(18(3)4)33(42)45-28(20(7)8)31(40)37(14)26(23(12)17-2)35(44)47-29(21(9)10)32(41)38(25)15/h18-29H,16-17H2,1-15H3/t22-,23-,24-,25-,26-,27+,28+,29+/m0/s1

InChI key

OWUREPXBPJFMOK-CIRFPNLUSA-N

Biochem/physiol Actions

Enniatins are a group of cyclohexadepsipeptide mycotoxins produced by Gnomonia errabuda and several Fusaria species, with phytotoxic, antibiotic, and insecticidal activities. Enniatins function as ionophors by their incorporation into the cellular membrane to form dimeric structures that transport monovalent ions across the membrane (especially the mitochondrial membranes) affecting oxidative phosphorylation uncoupling. It has been demonstrated that enniatins have a cytotoxic effect on human cancer cells. Furthermore, incubation of H4IIE hepatoma cells with enniatins strongly diminished phosphorylation of the ERK (p44/p42).
Enniatins are cyclohexadepsipeptide mycotoxins that have phytotoxic, antibiotic, and insecticidal activities and function as ionophors.


pictograms

Skull and crossbones

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library