Skip to Content
Merck

E5003

Sigma-Aldrich® micro pH combination electrode, glass body

Ferrule-pin connector, electrode L 132 mm

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NB.79
UNSPSC Code:
41115603
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


electrode L

132 mm

stem O.D.

4 mm

General description

Designed with a Refillable Ag/AgCl reference half cell. These electrodes feature ultra thin insertion stems and miniature pH bulbs for measurement of the smallest of samples. Depends on the length, the extra long insertion stem allows this combination electrode to measure samples upto the bottom of 7 in. long NMR tubes.

Sturdy and versatile probe for measuring small samples in cuvets and small lab glassware 0-14 pH.

Attention: These electrodes come with a little white plastic transport cap attached to the end, please remove before use.

Temperature Range:
  • -5 to 100°C (for continuous use)
  • -5 to 110°C (for intermittent use)

Other Notes

Help in selecting the correct electrode can be viewed at Electrode Selection Guide
Hints on Electrode Maintenance

Legal Information

SIGMA-ALDRICH is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content





Global Trade Item Number

SKUGTIN
E5003-1EA04061826761762