CLS432024
Corning® CoolBox™, XT System
workstation, single capacity, pink
Synonym(s):
Corning Coolcell accessory
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41123300
NACRES:
NB.12
Recommended Products
feature
workstation, single capacity
packaging
pkg of 1
manufacturer/tradename
Corning 432024
size
14.0 cm × 9.4 cm × 9.9 cm, Internal Dimensions
20.0 cm × 16.0 cm × 15.0 cm, External Dimensions
color
pink
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Corning CoolBox XT systems are bench top cooling workstations that provide sample cooling or freezing without ice, electricity or batteries. CoolBox systems are versatile and accommodate a variety of sample formats and temperatures. The modular design enables the use of CoolRack and CoolSink thermo-conductive sample modules to hold microfuge tubes, cryogenic vials, PCR tubes or plates, assay plates and more.
- Consists of a base, cooling cartridge/core and a lid
- Includes a magnetized collar and a 1° to 8°C temperature indicator for visual assurance of proper temperature
- To use with any 30 place Corning CoolRack Tubes Module
- The internal cooling or freezing cores provide 0.5° to 4°C for over 10 hours with lid open and over 16 hours with lid closed
- CoolBox systems may also be used with dry ice to create a compact, portable, snap freezing workstation for bacteria, virus or proteins.
Features and Benefits
Cooling with Lid Closed: 16 Hours
Cooling with Lid Open: 10 Hours
Freezing with Lid Closed: 8 Hours
Freezing with Lid Open: 5 Hours
Cooling with Lid Open: 10 Hours
Freezing with Lid Closed: 8 Hours
Freezing with Lid Open: 5 Hours
Legal Information
CoolBox is a trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service