CLS432010
Corning® CoolCell™ SV2 Freezing Container
for 12 x 2 mL serum vials
Synonym(s):
Corning Coolcell accessory
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
foamed PE
feature
Holds 12 x 2 mL serum vials
for 12 x 2 mL serum vials
packaging
pkg of 1
manufacturer/tradename
Corning 432010
technique(s)
cryopreservation: suitable
H × diam.
10.5 cm × 13.9 cm , 14.7 mm well diameter
Looking for similar products? Visit Product Comparison Guide
General description
Corning CoolCell alcohol-free cell freezing containers ensure standardized controlled-rate -1°C/minute cell freezing in a -80°C freezer - without alcohol or any fluids. Proven for use with a variety of cell types including stem cells, primary cells, PBMC cell lines, insect cells, yeast and others. The patentpending CoolCell technology utilizes a thermo-conductive alloy core and highly-insulative outer material to control the rate of heat removal and provide reproducible cell cryopreservation. CoolCell units are easy to use and deliver comparable results to expensive programmable freezers.
- For 12 standard 2.0 mL injectable ampules.
- Radially symmetric for uniform freezing of injectable ampules.
- Easy open lid.
- Exposed vial tops when lid is open for quick, organized removal of frozen samples.
Legal Information
CoolCell is a trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service