CLS3904BC
Corning® 96 Well Microplate With Generic Barcode
flat bottom clear, black polystyrene, Tissue Culture (TC)-treated surface, bag of 20 ×, sterile, lid
Synonym(s):
96 multiwell plate, 96 well barcode plate, 96 well microplate, 96 well microtiter plate, 96 well plate, 96 well plate with generic barcode
About This Item
flat bottom black polystyrene wells
flat bottom clear
polystyrene
Recommended Products
material
black polystyrene
flat bottom black polystyrene wells
flat bottom clear
polystyrene
sterility
sterile
feature
lid
skirt
plate format: 96 well standard
packaging
pack of 10
case of 50
bag of 20 ×
manufacturer/tradename
Corning 3904BC
maximum volume
320 μL
size
96 wells
working volume
200 μL
color
black
suitability
suitable for (cell culture)
binding type
Tissue Culture (TC)-treated surface
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
- Corning® 96 well microplate with Generic Barcode feature clear flat well bottom for direct microscopic viewing.
- Black polystyrene has low background fluorescence and minimizes light scattering.
- TC-treated for optimal cell attachment.
- Nonreversable lids with condensation ring to reduce contamination.
- Bottoms are 60% thinner than conventional polystyrene plates.
- Sterilized by gamma radiation and nonpyrogenic.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service