Skip to Content
CLS3333
Corning® CellSTACK® accessories
sterile, 6.4 mm I.D. tubing, 70 cm length, male MPC coupling with female end cap, cap (fill), case of 4
Synonym(s):
CellSTACK Accessories, Corning Accessories, Corning CellSTACK Accessories
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NB.13
UNSPSC Code:
41122104
Material:
cap (fill)
Sterility:
sterile
Manufacturer/tradename:
Corning 3333
Packaging:
case of 4
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
material
cap (fill)
sterility
sterile
packaging
case of 4
manufacturer/tradename
Corning 3333
General description
Available for filling, venting, stacking and manipulation.
Features and Benefits
  • Consists of 1/4 flexible tubing that is heat sealable and chemically resistant
  • Threaded cap made up of polyethylene
  • Gamma irradiation sterilized
Legal Information
CellSTACK is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.
Still not finding the right product?
Explore all of our products under Corning® CellSTACK® accessories
Choose from one of the most recent versions:
Certificates of Analysis (COA)