CLS3238
Corning® CellCube® Tubing Adaptor
3/8”ID Thermoelastic Tubing, Aseptic Connector to Male MPC, sterile
Synonym(s):
flask accessory
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
41122104
NACRES:
NB.13
Recommended Products
material
polystyrene
tubing (3/8”ID Thermoelastic)
sterility
sterile
packaging
pkg of 4 ea
manufacturer/tradename
Corning 3238
compatibility
for use with Corning HYPERStack 36-layer cell culture vessel
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
- CellCube Adaptor
- Aseptic Connector to Male MPC
Application
Adherent Cell Scale Up
Legal Information
CELLCUBE is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service