Skip to Content
Merck

CLS2016

Corning® cap inserts for cryogenic vials

blue

Synonym(s):

Corning cap

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NB.13
UNSPSC Code:
41121704
Color:
blue
Sterility:
non-sterile
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


sterility

non-sterile

manufacturer/tradename

Corning 2016

color

blue

General description

  • Cap inserts provide color coding for easy sample identification
  • Inserts are packaged in resealable bags
  • Cap inserts fit all Corning cryogenic vials except Corning No. 430289

Legal Information

Corning is a registered trademark of Corning, Inc.


Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
CLS2016-500EA04061833878156