Skip to Content

CLS2015

Corning® cap inserts for cryogenic vials

white

Synonym(s):

cryovials

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NB.13
UNSPSC Code:
41121704
Color:
white
Sterility:
non-sterile
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


sterility

non-sterile

manufacturer/tradename

Corning 2015

color

white

General description

  • Cap inserts provide color coding for easy sample identification
  • Inserts are packaged in resealable bags
  • Cap inserts fit all Corning cryogenic vials except Corning No. 430289

Features and Benefits

  • Corning® cap inserts for cryogenic vials are made of Virgin Polypropylene.
  • White caps inserts provide color-coding for easy sample identification.
  • Nonsterile.

Legal Information

Corning is a registered trademark of Corning, Inc.


Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library