Skip to Content
Merck
All Photos(1)

Key Documents

NB69

99072802, human brain, Not specified

Synonym(s):

NB 69, NB-69

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

Product Name

NB69, 99072802

biological source

human brain

growth mode

Semi-adherent

karyotype

Not specified

morphology

Not specified

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human neuroblastoma (Stage III)

Cell Line Description

Sample was isolated from a 16 month old male

DNA Profile

STR-PCR Data: Amelogenin: X,Y
CSF1PO: 11,12
D13S317: 8
D16S539: 9,11
D5S818: 11,12
D7S820: 10
THO1: 8
TPOX: 11
vWA: 17,18

Culture Medium

RPMI 1640 + 2mM Glutamine + 15% Foetal Bovine Serum (FBS).

Subculture Routine

Cells can grow in aggregates as well as attached. Split sub-confluent cultures 1:4 using 0.25% tryps

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service