Skip to Content
Merck
All Photos(1)

Key Documents

QT 6

93120831, quail muscle, Spindle shaped/round

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

Product Name

QT 6, 93120831

biological source

quail muscle

growth mode

Adherent

karyotype

Not specified

morphology

Spindle shaped/round

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Quail Japanese fibrosarcoma

Cell Line Description

Derived from methylcholanthrene-induced fibrosarcoma of Japanese quail. The cells are susceptible to several viruses including sub group A avian sarcoma, avian encephalomyelitis, Marek′s disease herpes virus and rabies virus.

Application

Virus studies

Culture Medium

Ham′s F10 + 2mM Glutamine + 10% Tryptose Phosphate Broth + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:2 to 1:4 i.e. seeding at 2-4x10,000 cells/cm2 using 0.25% trypsin/EDTA; 5% CO2; 37°C; subculture at about 75% confluency; cells can be dislodged by tapping flask against the palm of hand; use of 0.5% polyvinylpyrrol

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service