Skip to Content
Merck
All Photos(1)

Key Documents

TF1 Cell Line human

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA, 93022307, human blood, Not specified

Synonym(s):

Human TF1 Cells, TF1 Cell Culture

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

biological source

human blood

packaging

tube of 5 μg 93022307-DNA-5UG
pkg of vial of cells 93022307-1VL

growth mode

Suspension

karyotype

Carries homogeneous chromosomal abnormality (54,X)

morphology

Not specified

products

Not specified

receptors

Interleukin 3 (IL-3) and Granulocyte Macrophage Colony Stimulating Factor (GM-CSF)

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human erythroleukaemia

Cell Line Description

TF-1 have been derived from a patient with erythroleukaemia. They show complete growth dependency on GM-CSF or IL-3, and carry a homogeneous chromosomal abnormality (54,X). Constitutive expression of globin genes indicate commitment of TF-1 to the erythroid lineage, although glycophorin A or carbonyl anhydrase I are not expressed. On incubation with hemin or delta-aminolevulinic acid haemoglobin synthesis was induced, whereas TPA induced differentiation into macrophage-like cells. Erythroprotein also sustains short-term growth, but does not induce differentiation.

Application

TF1 cell line has been used-
  • for tumorigenic studies
  • to study the effect of granulocyte-macrophage colony-stimulating factor (GM-CSF) and interleukin-3 (IL-3) on proliferation and TNFR2 expression in TF1 cells
  • in a cell-based proliferation assay to study the biological activities of anti-IL-6R Nanobody ALX-0061
  • to study the induction of STAT5 phosphorylation by IL-3

DNA Profile

STR-PCR Data: Amelogenin: X,Y
CSF1PO: 13
D13S317: 8,9
D16S539: 9,12
D5S818: 13
D7S820: 12
THO1: 7,9
TPOX: 8
vWA: 15,17

Culture Medium

RPMI 1640 + 2mM Glutamine + 1% Sodium Pyruvate (NaP) + 2-5ng/ml Human Granulocyte Macrophage Colony Stimulating Factor (Human GM-CSF) + 10% Foetal Bovine Serum (FBS). It is recommended to add fresh GM-CSF every 48 hours.

Subculture Routine

Maintain cultures between 2-9x100,000 cells/ml; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service