product name
L1210, 87092804
biological source
mouse blood
growth mode
Suspension
karyotype
2n=40, diploid
morphology
Lymphoblast
products
Not specified
receptors
Not specified
technique(s)
cell culture | mammalian: suitable
relevant disease(s)
cancer
shipped in
dry ice
storage temp.
−196°C
Cell Line Origin
Mouse DBA/2 lymphocytic leukaemia
Cell Line Description
Established from a tumour induced in an 8 month old female 234 subline 212 of a DBA mouse. Orginally subcultured in DBA/2 mice it was adapted to suspension culture. The line has been extensively used in cytotoxicity assays. Develops massive tumours in nude mice after a latent period of 7-10 days
Culture Medium
Fischer′s medium + 2mM Glutamine + 10% Horse Serum (HS).
Subculture Routine
Maintain cultures between 5x10,000 to 8x100,000 cells/ml; 5% CO2; 37°C.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service