KARPAS 299
6072604, human lymph, Lymphoblastoid
Synonym(s):
K299 Cells, KARPAS299 Cells
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106514
Recommended Products
Product Name
KARPAS 299, 06072604
biological source
human lymph
packaging
tube of 5 μg 06072604-DNA-5UG
pkg of vial of cells 06072604-1VL
growth mode
Suspension
karyotype
Hypodiploid
morphology
Lymphoblastoid
products
Not specified
receptors
Not specified
technique(s)
cell culture | mammalian: suitable
relevant disease(s)
cancer
shipped in
dry ice
storage temp.
−196°C
Related Categories
Cell Line Origin
Human Non-Hodgkin′s Ki-positive Large Cell Lymphoma
Cell Line Description
A human T cell lymphoma cell line established from the peripheral blood of a 25-year-old man with T cell non-Hodgkin′s lymphoma in 1986.
DNA Profile
STR-PCR Data: Amelogenin: X,Y
CSF1PO: 10,11
D13S317: 8,12
D16S539: 12
D5S818: 11,12
D7S820: 10,11
THO1: 7,8.3
TPOX: 8
vWA: 17,19
CSF1PO: 10,11
D13S317: 8,12
D16S539: 12
D5S818: 11,12
D7S820: 10,11
THO1: 7,8.3
TPOX: 8
vWA: 17,19
Culture Medium
RPMI 1640 + 2mM Glutamine + 20% Foetal Bovine Serum (FBS). For slower growth use 10% or 5% FBS.
Subculture Routine
Maintain cultures between 0.5 - 2 x1000,000 cells/ml; 5% CO2; 37°C. Doubling time approximately 30 hours. Split saturated culture 1:3 every 2-3 days. Round to polygonal cells growing singly or in small clusters in suspension.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Disclaimer
This cell line has special release conditions: Commercial organisations are required to complete the ′Cell Line Release Authorisation for Research Use in Commercial Organisations′ release conditions form.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service