Skip to Content
Merck
All Photos(3)

Key Documents

C3787

Sigma-Aldrich

Chloramphenicol succinate sodium salt

≥80% (HPLC)

Synonym(s):

Chloramphenicol α-succinate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C15H15Cl2N2O8Na
CAS Number:
Molecular Weight:
445.18
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.85

Assay

≥80% (HPLC)

form

solid

solubility

H2O: 50 mg/mL

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria

Mode of action

protein synthesis | interferes

SMILES string

[Na].O[C@@H]([C@@H](COC(=O)CCC(O)=O)NC(=O)C(Cl)Cl)c1ccc(cc1)N(=O)=O

InChI

1S/C15H16Cl2N2O8.Na.H/c16-14(17)15(24)18-10(7-27-12(22)6-5-11(20)21)13(23)8-1-3-9(4-2-8)19(25)26;;/h1-4,10,13-14,23H,5-7H2,(H,18,24)(H,20,21);;/t10-,13-;;/m1../s1

InChI key

RJOAHMNSYANTPN-OWVUFADGSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: phenicole

Application

Chloramphenicol is often used for bacterial selection in molecular biology applications at 10-20 μg/mL and as a selection agent for transformed cells containing chloramphenicol reistance genes.

Biochem/physiol Actions

Chloramphenicol is a synthetic antibiotic, isolated from strains of Streptomyces venezuelae. It inhibits bacterial protein synthesis by blocking the peptidyl transferase step by binding to the 50S ribosomal subunit and preventing attachment of aminoacyl tRNA to the ribosome. It also inhibits mitochondrial and chloroplast protein synthesis and ribosomal formation of (p)ppGpp, de-pressing rRNA transcription.

Mode of Resistance: Use of chloramphenicol acetyltransferase will acetylate the product and inactivate it.

Antimicrobial Spectrum: This is a broad spectrum antibiotic against gram-positive and gram-negative bacteria, and is used mainly for ophthalmic and veterinary purposes.

Caution

Stock solutions should be stored at 2-8°C and are stable at 37°C for 5 days. Aqueous solutions are neutral and stable over a wide pH range, with 50% hydrolysis occurring after 290 days. Use of a borax buffered solution reduces this number to 14%. Solutions should be protected from light as photochemical decomposition results in a yellowing of the solution. Heating aqueous solutions at 115°C for 30 minutes results in a 10% loss of chloramphenicol.

Preparation Note

Stock solutions can be prepared directly in the vial at any recommended concentration. A solution at 50 mg/mL in ethanol yields a clear, very faint, yellow solution. Degradation of chloramphenicol in aqueous solution is catalyzed by general acids and bases. This rate of degradation is independent of the ionic strength and pH.

Other Notes

Keep container tightly closed in a dry and well-ventilated place.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Studies on the mechanism of action of chloramphenicol. I. The conformation of chlioramphenicol in solution.
O JARDETZKY
The Journal of biological chemistry, 238, 2498-2508 (1963-07-01)
G C Allen et al.
Applied and environmental microbiology, 56(4), 1025-1032 (1990-04-01)
Succinate- or oxygen-limited continuous cultures were used to study the influences of different concentrations of dissolved oxygen and ammonia on the growth, respiration, and polypeptide patterns of Bradyrhizobium sp. (Arachis) strain 3G4b20. During succinate-limited growth, molar growth yields on succinate
Karin Przyklenk et al.
Autophagy, 7(4), 432-433 (2010-12-29)
There is no question that necrosis and apoptosis contribute to cardiomyocyte death in the setting of myocardial ischemia-reperfusion. Indeed, considerable effort and resources have been invested in the development of novel therapies aimed at attenuating necrotic and apoptotic cell death
Simultaneous measurement of chloramphenicol sodium succinate and chloramphenicol in presence of furosemide in plasma and urine.
M C Nahata
Journal of clinical pharmacy and therapeutics, 18(4), 301-301 (1993-08-01)
John A Turton et al.
International journal of experimental pathology, 87(2), 101-112 (2006-04-21)
Chloramphenicol (CAP) is haemotoxic in man, inducing two types of toxicity. First, a dose-related, reversible anaemia with reticulocytopenia, sometimes seen in conjunction with leucopenia and thrombocytopenia; this form of toxicity develops during drug treatment. The second haemotoxicity is aplastic anaemia

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service