BMSIPD960038781
BeadBlaster™ 96 Ceramic grinding bar, 45° angled ends
bead diam. × L 3/8 × 7/8 , fits 50 mL tubes
All Photos(1)
About This Item
UNSPSC Code:
41100000
NACRES:
NA.82
Recommended Products
material
ceramic
packaging
pack of 100
manufacturer/tradename
Benchmark Scientific IPD960038781
bead diam. × L
3/8 × 7/8 , fits 50 mL tubes
compatibility
BeadBlaster™ 96
Related Categories
General description
Ceramic grinding media is suitable for applications where highly efficient grinding must be paired with low grinding media abrasion. Non-ferrous grinding is absolutely essential in many processes, for instance in medical or pharmaceutical applications, in order to prevent undesired impurities in the grinding materials used. Ceramic grinding media are also suited for use in processes where mixing and dispersing effects play an important role.
Features and Benefits
- low abrasion with high efficiency grinding
- low impurity transfer
Legal Information
BeadBlaster is a trademark of Benchmark Scientific, Inc.